(2E)-1-(2-Methoxyphenyl)-3-[2-(trifluoromethyl)phenyl]-2-propen-1-one ≥97%

Supplier: AOBChem USA

Warning

302980-5G 302980-25G 302980-1G 302980-500MG
77632-574EA 1441 USD
77632-574 77654-840 77641-858 77649-918
(2E)-1-(2-Methoxyphenyl)-3-[2-(trifluoromethyl)phenyl]-2-propen-1-one ≥97%
(2E)-1-(2-Methoxyphenyl)-3-[2-(trifluoromethyl)phenyl]-2-propen-1-one

Precaución: For Research Use Only

Formula: 306.28 g/mol
MW: C₁₇H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD30186783
CAS Number: 1309371-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR