3-(2-Pyridinyl)phenol ≥95%

Supplier: AOBChem USA

Warning

302869-5G 302869-1G 302869-500MG 302869-25G
77632-572EA 849 USD
77632-572 77637-904 77652-616 77654-838
3-(2-Pyridinyl)phenol ≥95%
3-(2-Pyridinyl)phenol

Precaución: For Research Use Only

Formula: 171.2 g/mol
MW: C₁₁H₉NO
Storage Temperature: Ambient
MDL Number: MFCD08705744
CAS Number: 98061-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR