Methyl 4'-nitro-[1,1'-biphenyl]-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

302810-5G 302810-1G 302810-25G
77634-690EA 232 USD
77634-690 77637-902 77654-834
Methyl 4'-nitro-[1,1'-biphenyl]-3-carboxylate ≥97%
Methyl 4'-nitro-[1,1'-biphenyl]-3-carboxylate

Precaución: For Research Use Only

Formula: C₁₄H₁₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD20529487
CAS Number: 107558-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR