3-(2-Methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

30281-5G 30281-25G 30281-1G 30281-500MG
77632-570EA 892 USD
77632-570 77654-836 77641-856 77645-860
3-(2-Methylphenyl)pyridine ≥97%
3-(2-Methylphenyl)pyridine

Precaución: For Research Use Only

Formula: C₁₂H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD06801824
CAS Number: 90395-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR