2-Bromo-4-fluoro-3-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

302365-1G 302365-500MG 302365-5G
77634-686EA 807 USD
77634-686 77652-614 77658-574
2-Bromo-4-fluoro-3-methylpyridine ≥95%
2-Bromo-4-fluoro-3-methylpyridine

Precaución: For Research Use Only

Formula: C₆H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD16609961
CAS Number: 1227599-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR