1-Bromo-4-chloro-2-((3-methylbenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

302304-5G 302304-25G 302304-1G 302304-500MG
77628-482EA 1484.23 USD
77628-482 77658-572 77641-852 77645-856
1-Bromo-4-chloro-2-((3-methylbenzyl)oxy)benzene ≥97%
1-Bromo-4-chloro-2-((3-methylbenzyl)oxy)benzene

Precaución: For Research Use Only

Formula: 311.6014 g/mol
MW: C₁₄H₁₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD34891668
CAS Number: 2144145-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR