2-Isopropoxy-5-nitropyridine ≥95%

Supplier: AOBChem USA

Warning

30218-5G 30218-1G 30218-25G 30218-500MG
77632-566EA 931 USD
77632-566 77637-898 77658-570 77649-914
2-Isopropoxy-5-nitropyridine ≥95%
2-Isopropoxy-5-nitropyridine

Precaución: For Research Use Only

Formula: C₈H₁₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD07374880
CAS Number: 24903-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR