2-Bromo-1,3-dimethyl-5-(methylthio)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
301932-5G
301932-25G
301932-1G
301932-500MG
77632-562EA
1200
USD
77632-562
77658-568
77641-850
77649-910
2-Bromo-1,3-dimethyl-5-(methylthio)benzene ≥97%
2-Bromo-1,3-dimethyl-5-(methylthio)benzene
Precaución: For Research Use Only
Formula:
231.15 g/mol MW: C₉H₁₁BrS Storage Temperature: Ambient |
MDL Number:
MFCD32691134 CAS Number: 1267535-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...