Methyl 6-phenylnicotinate ≥95%

Supplier: AOBChem USA

Warning

30168-5G 30168-1G 30168-25G 30168-500MG
77634-684EA 610 USD
77634-684 77637-892 77654-826 77649-906
Methyl 6-phenylnicotinate ≥95%
Methyl 6-phenylnicotinate

Precaución: For Research Use Only

Formula: C₁₃H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD07367320
CAS Number: 4634-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR