Benzoic acid, 4-phenoxy-, ethyl ester ≥97%

Supplier: AOBChem USA

Warning

301281-5G 301281-1G 301281-25G 301281-500MG
77632-558EA 732 USD
77632-558 77637-890 77658-562 77649-904
Benzoic acid, 4-phenoxy-, ethyl ester ≥97%
Benzoic acid, 4-phenoxy-, ethyl ester

Precaución: For Research Use Only

Formula: 242.27 g/mol
MW: C₁₅H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00496636
CAS Number: 31994-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR