5-Chloro-2-isopropoxy-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

30087-5G 30087-1G 30087-500MG
77628-480EA 1484.23 USD
77628-480 77641-846 77645-854
5-Chloro-2-isopropoxy-3-iodopyridine ≥97%
5-Chloro-2-isopropoxy-3-iodopyridine

Precaución: For Research Use Only

Formula: 297.521 g/mol
MW: C₈H₉ClINO
Storage Temperature: Ambient
MDL Number: MFCD29088134
CAS Number: 1826109-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR