1,2-Dihydro-5-(trimethylsilyl)acenaphthylene ≥95%

Supplier: AOBChem USA

Warning

299816-5G 299816-1G 299816-500MG
77654-820EA 2140 USD
77654-820 77645-848 77649-896
1,2-Dihydro-5-(trimethylsilyl)acenaphthylene ≥95%
1,2-Dihydro-5-(trimethylsilyl)acenaphthylene

Precaución: For Research Use Only

Formula: C₁₅H₁₈Si
Storage Temperature: Ambient
MDL Number: MFCD27965448
CAS Number: 60989-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR