1-Chloro-2,4-difluoro-5-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

29963-5G 29963-25G 29963-1G 29963-500MG
77632-554EA 940 USD
77632-554 77654-812 77641-836 77649-894
1-Chloro-2,4-difluoro-5-iodobenzene ≥95%
1-Chloro-2,4-difluoro-5-iodobenzene

Precaución: For Research Use Only

Formula: 274.434 g/mol
MW: C₆H₂ClF₂I
Storage Temperature: Ambient
MDL Number: MFCD18415604
CAS Number: 333447-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR