3'-(Dimethylamino)acetophenone ≥97%

Supplier: AOBChem USA

Warning

299235-5G 299235-25G
77634-670EA 98 USD
77634-670 77654-810
3'-(Dimethylamino)acetophenone ≥97%
3'-(Dimethylamino)acetophenone

Precaución: For Research Use Only

Formula: C₁₀H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00059343
CAS Number: 18992-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR