5-Bromo-2-((4-(tert-butyl)benzyl)thio)-4-methylpyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
299230-5G
299230-25G
299230-1G
299230-500MG
77628-470EA
1484.23
USD
77628-470
77635-616
77637-880
77649-892
5-Bromo-2-((4-(tert-butyl)benzyl)thio)-4-methylpyridine ≥95%
5-Bromo-2-((4-(tert-butyl)benzyl)thio)-4-methylpyridine
Precaución: For Research Use Only
Formula:
350.3164 g/mol MW: C₁₇H₂₀BrNS Storage Temperature: Ambient |
MDL Number:
MFCD34890838 CAS Number: 2969392-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...