5-Bromo-2-((4-(tert-butyl)benzyl)thio)pyridine ≥97%

Supplier: AOBChem USA

Warning

299229-5G 299229-25G 299229-1G 299229-500MG
77628-468EA 1484.23 USD
77628-468 77635-614 77637-878 77645-844
5-Bromo-2-((4-(tert-butyl)benzyl)thio)pyridine ≥97%
5-Bromo-2-((4-(tert-butyl)benzyl)thio)pyridine

Precaución: For Research Use Only

Formula: 336.2898 g/mol
MW: C₁₆H₁₈BrNS
Storage Temperature: Ambient
MDL Number: MFCD34890837
CAS Number: 2969376-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR