Methyl 4-chloro-2-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

29918-5G 29918-1G 29918-500MG
77628-466EA 1484.23 USD
77628-466 77641-834 77645-840
Methyl 4-chloro-2-fluoro-5-methylbenzoate ≥97%
Methyl 4-chloro-2-fluoro-5-methylbenzoate

Precaución: For Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28363898
CAS Number: 1192547-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR