4-(Naphthalen-1-yl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

299164-5G 299164-1G 299164-500MG 299164-25G
77632-550EA 855 USD
77632-550 77637-874 77652-610 77658-554
4-(Naphthalen-1-yl)benzoic acid ≥95%
4-(Naphthalen-1-yl)benzoic acid

Precaución: For Research Use Only

Formula: 248.2759 g/mol
MW: C₁₇H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04117438
CAS Number: 106359-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR