(4-Bromo-3-chloro-2-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

29911-5G 29911-1G 29911-25G 29911-500MG
77632-548EA 635 USD
77632-548 77637-872 77654-808 77645-838
(4-Bromo-3-chloro-2-methylphenyl)methanol ≥95%
(4-Bromo-3-chloro-2-methylphenyl)methanol

Precaución: For Research Use Only

Formula: 235.506 g/mol
MW: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD29036609
CAS Number: 1809168-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR