Ethyl 2,2-difluoro-2-(2-methoxyphenyl)acetate ≥97%

Supplier: AOBChem USA

Warning

298630-5G 298630-25G
77634-666EA 145 USD
77634-666 77658-550
Ethyl 2,2-difluoro-2-(2-methoxyphenyl)acetate ≥97%
Ethyl 2,2-difluoro-2-(2-methoxyphenyl)acetate

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026027
CAS Number: 1150164-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR