3-Methyl-4-(methylthio)phenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29734-5G 29734-25G 29734-1G 29734-500MG
77632-542EA 760 USD
77632-542 77658-546 77645-830 77645-832
3-Methyl-4-(methylthio)phenylboronic acid, pinacol ester ≥97%
3-Methyl-4-(methylthio)phenylboronic acid, pinacol ester

Precaución: For Research Use Only

Formula: C₁₄H₂₁BO₂S
Storage Temperature: Ambient
MDL Number: MFCD29036606
CAS Number: 2121514-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR