α-Ethyl-2-methyl-5-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

297285-5G 297285-25G 297285-1G 297285-500MG
77628-464EA 1639.76 USD
77628-464 77635-612 77637-866 77649-886
α-Ethyl-2-methyl-5-(trifluoromethyl)benzenemethanol ≥95%
α-Ethyl-2-methyl-5-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 218.22 g/mol
MW: C₁₁H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD34148398
CAS Number: 2151082-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR