1-(Benzyloxy)-2-bromo-4,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

29685-5G 29685-25G 29685-1G 29685-500MG
77632-538EA 656 USD
77632-538 77654-800 77641-828 77649-884
1-(Benzyloxy)-2-bromo-4,5-dimethylbenzene ≥95%
1-(Benzyloxy)-2-bromo-4,5-dimethylbenzene

Precaución: For Research Use Only

Formula: 291.183 g/mol
MW: C₁₅H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD29036599
CAS Number: 1809168-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR