4,5-Dimethyl-2-ethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

29683-5G 29683-1G 29683-500MG
77628-458EA 1777.78 USD
77628-458 77637-858 77649-882
4,5-Dimethyl-2-ethoxyphenylboronic acid ≥97%
4,5-Dimethyl-2-ethoxyphenylboronic acid

Precaución: For Research Use Only

Formula: 194.035 g/mol
MW: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD29036597
CAS Number: 2121514-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR