4-Bromo-2-chloro-1-(2-chlorophenoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

296490-5G 296490-25G 296490-1G 296490-500MG
77628-456EA 1725.25 USD
77628-456 77635-610 77641-824 77645-820
4-Bromo-2-chloro-1-(2-chlorophenoxy)benzene ≥97%
4-Bromo-2-chloro-1-(2-chlorophenoxy)benzene

Precaución: For Research Use Only

Formula: 317.9933 g/mol
MW: C₁₂H₇BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD34889893
CAS Number: 2969462-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR