4-Bromo-2-chloro-1-(o-tolyloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

296484-5G 296484-25G 296484-1G 296484-500MG
77628-454EA 1484.23 USD
77628-454 77635-608 77641-822 77649-880
4-Bromo-2-chloro-1-(o-tolyloxy)benzene ≥97%
4-Bromo-2-chloro-1-(o-tolyloxy)benzene

Precaución: For Research Use Only

Formula: C₁₃H₁₀BrClO
Storage Temperature: Ambient
MDL Number: MFCD34889889
CAS Number: 2969462-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR