3-(2-Methoxyethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

296225-5G 296225-25G 296225-1G 296225-500MG
77634-658EA 339 USD
77634-658 77654-792 77641-814 77649-874
3-(2-Methoxyethoxy)benzoic acid ≥95%
3-(2-Methoxyethoxy)benzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD09036534
CAS Number: 152808-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR