4-[2-(4-Bromophenoxy)ethyl]morpholine ≥97%

Supplier: AOBChem USA

Warning

29619-5G 29619-25G 29619-1G
77634-656EA 226 USD
77634-656 77658-534 77641-812
4-[2-(4-Bromophenoxy)ethyl]morpholine ≥97%
4-[2-(4-Bromophenoxy)ethyl]morpholine

Precaución: For Research Use Only

Formula: C₁₂H₁₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD01308572
CAS Number: 836-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR