3-(3-Thienyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

296042-5G 296042-1G 296042-25G 296042-500MG
77632-524EA 990 USD
77632-524 77637-850 77654-790 77649-872
3-(3-Thienyl)benzonitrile ≥95%
3-(3-Thienyl)benzonitrile

Precaución: For Research Use Only

Formula: C₁₁H₇NS
Storage Temperature: Ambient
MDL Number: MFCD06409245
CAS Number: 870703-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR