2-[3.4-Difluoro-2-(trimethylsilyl)phenyl]-1,3-dioxolane ≥97%
Supplier: AOBChem USA
|
Warning
|
29581-5G
29581-1G
29581-25G
29581-500MG
77634-654EA
256
USD
77634-654
77637-848
77658-530
77645-814
2-[3.4-Difluoro-2-(trimethylsilyl)phenyl]-1,3-dioxolane ≥97%
2-[3.4-Difluoro-2-(trimethylsilyl)phenyl]-1,3-dioxolane
Precaución: For Research Use Only
Formula:
258.337 g/mol MW: C₁₂H₁₆F₂O₂Si Storage Temperature: Ambient |
MDL Number:
MFCD29036595 CAS Number: 1809168-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...