4-(Cyclopropylmethoxy)-2-fluorobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

295777-5G 295777-25G 295777-1G 295777-500MG
77632-522EA 736 USD
77632-522 77654-788 77641-810 77649-870
4-(Cyclopropylmethoxy)-2-fluorobenzonitrile ≥95%
4-(Cyclopropylmethoxy)-2-fluorobenzonitrile

Precaución: For Research Use Only

Formula: C₁₁H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD23426444
CAS Number: 1369882-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR