3,5-Dimethyl-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29528-5G 29528-25G 29528-1G 29528-500MG
77634-650EA 321 USD
77634-650 77658-526 77641-808 77645-812
3,5-Dimethyl-4-isopropoxybenzoic acid ≥97%
3,5-Dimethyl-4-isopropoxybenzoic acid

Precaución: For Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD11188408
CAS Number: 1014691-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR