3-(1,1-Dimethylethyl)-α-ethyl-4-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

294850-5G 294850-1G 294850-500MG
77654-786EA 2348 USD
77654-786 77641-806 77649-868
3-(1,1-Dimethylethyl)-α-ethyl-4-methoxybenzenemethanol ≥95%
3-(1,1-Dimethylethyl)-α-ethyl-4-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: 222.32 g/mol
MW: C₁₄H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD14530443
CAS Number: 35154-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR