(3-Bromobenzyl)(p-tolyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

294812-5G 294812-25G 294812-1G 294812-500MG
77632-520EA 1441 USD
77632-520 77658-524 77641-804 77649-866
(3-Bromobenzyl)(p-tolyl)sulfane ≥97%
(3-Bromobenzyl)(p-tolyl)sulfane

Precaución: For Research Use Only

Formula: 293.2220 g/mol
MW: C₁₄H₁₃BrS
Storage Temperature: Ambient
MDL Number: MFCD18262578
CAS Number: 1457871-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR