Ethyl 2-fluoro-3,6-dimethoxy-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

294507-5G 294507-1G 294507-500MG
77658-520EA 2314 USD
77658-520 77641-802 77645-810
Ethyl 2-fluoro-3,6-dimethoxy-α-oxobenzeneacetate ≥95%
Ethyl 2-fluoro-3,6-dimethoxy-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 256.23 g/mol
MW: C₁₂H₁₃FO₅
Storage Temperature: Ambient
MDL Number: MFCD34889378
CAS Number: 1898103-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR