1-[2,2'-Bithiophen]-5-yl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

294359-5G 294359-1G 294359-500MG
77628-450EA 1484.23 USD
77628-450 77641-800 77645-808
1-[2,2'-Bithiophen]-5-yl-1-propanone ≥95%
1-[2,2'-Bithiophen]-5-yl-1-propanone

Precaución: For Research Use Only

Formula: 222.33 g/mol
MW: C₁₁H₁₀OS₂
Storage Temperature: Ambient
MDL Number: MFCD04039750
CAS Number: 32358-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR