3-(3-Chlorophenyl)-1-propene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Allylchlorobenzene

293677-5G 293677-25G
77634-648EA 95 USD
77634-648 77654-782
3-(3-Chlorophenyl)-1-propene ≥97%
3-(3-Chlorophenyl)-1-propene

Precaución: For Research Use Only

Formula: C₉H₉Cl
MW: 152.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319545
CAS Number: 3840-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR