Methyl 4-cyclopropyl-2-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
29283-1G
29283-5G
29283-500MG
77637-840EA
519
USD
77637-840
77654-774
77645-802
Methyl 4-cyclopropyl-2-methylbenzoate ≥97%
Methyl 4-cyclopropyl-2-methylbenzoate
Precaución: For Research Use Only
Formula:
C₁₂H₁₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD27932149 CAS Number: 909698-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...