1-Bromo-3-methyl-5-((4-methylbenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

292671-5G 292671-25G 292671-1G 292671-500MG
77628-448EA 1484.23 USD
77628-448 77654-772 77641-792 77645-798
1-Bromo-3-methyl-5-((4-methylbenzyl)oxy)benzene ≥97%
1-Bromo-3-methyl-5-((4-methylbenzyl)oxy)benzene

Precaución: For Research Use Only

Formula: 291.183 g/mol
MW: C₁₅H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD34888932
CAS Number: 2969338-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR