1-Bromo-2-methyl-4-((3-methylbenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

292655-5G 292655-1G 292655-25G 292655-500MG
77628-446EA 1484.23 USD
77628-446 77637-838 77658-514 77649-860
1-Bromo-2-methyl-4-((3-methylbenzyl)oxy)benzene ≥97%
1-Bromo-2-methyl-4-((3-methylbenzyl)oxy)benzene

Precaución: For Research Use Only

Formula: 291.183 g/mol
MW: C₁₅H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD25884521
CAS Number: 1555527-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR