(1,2-Dihydroacenaphthylen-5-yl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

292620-5G 292620-25G 292620-1G 292620-500MG
77632-508EA 1441 USD
77632-508 77654-770 77641-790 77649-858
(1,2-Dihydroacenaphthylen-5-yl)(methyl)sulfane ≥95%
(1,2-Dihydroacenaphthylen-5-yl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 200.2993 g/mol
MW: C₁₃H₁₂S
Storage Temperature: Ambient
MDL Number: MFCD34888893
CAS Number: 19162-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR