1-Bromo-2-chloro-4-fluoro-3-(2-methoxyethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

292584-5G 292584-1G 292584-25G 292584-500MG
77628-444EA 1484.23 USD
77628-444 77637-836 77654-768 77645-796
1-Bromo-2-chloro-4-fluoro-3-(2-methoxyethoxy)benzene ≥97%
1-Bromo-2-chloro-4-fluoro-3-(2-methoxyethoxy)benzene

Precaución: For Research Use Only

Formula: 283.5219 g/mol
MW: C₉H₉BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD35169629
CAS Number: 3002468-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR