4-(4-Bromo-3-methylphenyl)morpholine ≥95%

Supplier: AOBChem USA

Warning

29257-5G 29257-25G 29257-1G 29257-500MG
77632-506EA 1181 USD
77632-506 77654-766 77641-788 77649-856
4-(4-Bromo-3-methylphenyl)morpholine ≥95%
4-(4-Bromo-3-methylphenyl)morpholine

Precaución: For Research Use Only

Formula: 256.139 g/mol
MW: C₁₁H₁₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD24133740
CAS Number: 1366131-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR