1-(4-Bromo-2-methylphenyl)piperidine ≥95%

Supplier: AOBChem USA

Warning

29254-5G 29254-1G 29254-25G 29254-500MG
77632-504EA 661 USD
77632-504 77637-834 77658-510 77645-794
1-(4-Bromo-2-methylphenyl)piperidine ≥95%
1-(4-Bromo-2-methylphenyl)piperidine

Precaución: For Research Use Only

Formula: 254.166 g/mol
MW: C₁₂H₁₆BrN
Storage Temperature: Ambient
MDL Number: MFCD20919961
CAS Number: 1020180-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR