4-Benzyloxy-2-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

29238-5G 29238-1G 29238-500MG
77628-442EA 1484.23 USD
77628-442 77641-786 77649-854
4-Benzyloxy-2-(trifluoromethoxy)benzoic acid ≥95%
4-Benzyloxy-2-(trifluoromethoxy)benzoic acid

Precaución: For Research Use Only

Formula: 312.241 g/mol
MW: C₁₅H₁₁F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD28806030
CAS Number: 1809161-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR