1-(3-Bromo-4-hydroxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3'-Bromo-4'-hydroxyacetophenone, 3-Bromo-4-hydroxyacetophenone

29222-5G 29222-25G
77634-644EA 65 USD
77634-644 77654-764
1-(3-Bromo-4-hydroxyphenyl)ethanone ≥95%
1-(3-Bromo-4-hydroxyphenyl)ethanone

Precaución: For Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03208974
CAS Number: 1836-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR