4-Methoxy-2-(trifluoromethoxy)phenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29196-5G 29196-1G 29196-25G 29196-500MG
77632-500EA 786 USD
77632-500 77637-830 77654-762 77645-790
4-Methoxy-2-(trifluoromethoxy)phenylboronic acid, pinacol ester ≥97%
4-Methoxy-2-(trifluoromethoxy)phenylboronic acid, pinacol ester

Precaución: For Research Use Only

Formula: C₁₄H₁₈BF₃O₄
Storage Temperature: Ambient
MDL Number: MFCD28806021
CAS Number: 2972608-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR