1-(2-Chloro-3,5-difluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

291889-5G 291889-25G 291889-1G 291889-500MG
77628-440EA 1484.23 USD
77628-440 77635-606 77641-784 77645-788
1-(2-Chloro-3,5-difluorophenyl)ethanone ≥95%
1-(2-Chloro-3,5-difluorophenyl)ethanone

Precaución: For Research Use Only

Formula: 190.575 g/mol
MW: C₈H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD28640126
CAS Number: 1557263-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR