2-(3,5-Difluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

290825-5G 290825-25G 290825-1G 290825-500MG
77632-496EA 967 USD
77632-496 77658-502 77641-782 77649-850
2-(3,5-Difluorophenyl)propan-2-ol ≥95%
2-(3,5-Difluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 172.1719 g/mol
MW: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD17222271
CAS Number: 1063733-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR