1-Benzyl-3-bromobenzene ≥97%

Supplier: AOBChem USA

Warning

290053-5G 290053-25G 290053-1G 290053-500MG
77632-492EA 1352 USD
77632-492 77654-754 77641-776 77649-846
1-Benzyl-3-bromobenzene ≥97%
1-Benzyl-3-bromobenzene

Precaución: For Research Use Only

Formula: C₁₃H₁₁Br
Storage Temperature: Ambient
MDL Number: MFCD18432375
CAS Number: 27798-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR